Assay Details
Target Gene Details
Entrez Gene ID: | 26400 |
Gene Name: | mitogen-activated protein kinase kinase 7 |
Gene Aliases: |
5930412N11Rik, JNKK 2, Jnkk2, MAPKK 7, MEK 7, Mapkk7, Mek7, Mkk7, Prkmk7, sek2 |
Location: |
Chr.8:4238740-4247897 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map2k7 | NM_001042557.2 | NP_001036022.1 | ||
| NM_001164172.1 | NP_001157644.1 | |||
| NM_001291777.1 | NP_001278706.1 | |||
| NM_001291778.1 | NP_001278707.1 | |||
| NM_001291783.1 | NP_001278712.1 | |||
| NM_011944.3 | NP_036074.2 | |||
| AB005654.1 | BAA24383.1 | |||
| AF022112.1 | AAC16274.1 | |||
| AF022113.1 | AAC16275.1 | |||
| AF060943.1 | AAD15819.1 | |||
| AF060944.1 | AAD15820.1 | |||
| AF060945.1 | AAD15821.1 | |||
| AF060946.1 | AAD15822.1 | |||
| AF060947.1 | AAD15823.1 | |||
| AK031137.1 | BAC27272.1 | |||
| AK165184.1 | BAE38066.1 | |||
| AK214564.1 | ||||
| BC070467.1 | AAH70467.1 | |||
| U74463.1 | AAC53364.1 | |||
| U74464.1 | AAC53365.1 | |||
| U93030.1 | AAB63447.1 |
More Information
Set Membership: |
|

Genomic Map