Assay Details
Target Gene Details
Entrez Gene ID: | 15368 |
Gene Name: | heme oxygenase 1 |
Gene Aliases: |
D8Wsu38e, HO-1, HO1, Hemox, Hmox, Hsp32 |
Location: |
Chr.8:75093618-75100593 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hmox1 | NM_010442.2 | NP_034572.1 | ||
| AK150934.1 | BAE29970.1 | |||
| AK151005.1 | BAE30028.1 | |||
| AK151012.1 | ||||
| AK151627.1 | ||||
| AK153420.1 | BAE31980.1 | |||
| AK153516.1 | BAE32059.1 | |||
| AK153566.1 | BAE32100.1 | |||
| AK154598.1 | BAE32701.1 | |||
| AK159521.1 | BAE35150.1 | |||
| AK159870.1 | BAE35443.1 | |||
| AK159959.1 | BAE35514.1 | |||
| AK169227.1 | BAE40996.1 | |||
| BC010757.1 | AAH10757.1 | |||
| BY319630.1 | ||||
| M33203.1 | AAA39872.1 | |||
| X13356.1 | CAA31732.1 |
More Information
Set Membership: |
|

Genomic Map