Assay Details
Target Gene Details
Entrez Gene ID: | 65972 |
Gene Name: | interferon gamma inducible protein 30 |
Gene Aliases: |
GILT, IP30 |
Location: |
Chr.8:70762773-70766663 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifi30 | NM_023065.3 | 5 | 552 | NP_075552.2 |
| AF290973.1 | 5 | 553 | AAG00598.1 | |
| AF309649.1 | 5 | 520 | AAG34681.1 | |
| AK077725.1 | 5 | 552 | ||
| AK156914.1 | 5 | 553 | BAE33895.1 | |
| BC054852.1 | 5 | 538 | AAH54852.1 | |
| BE951500.1 | 3 | 446 | ||
| BQ746455.1 | 5 | 508 |
More Information
Set Membership: |
|

Genomic Map