Assay Details
Target Gene Details
Entrez Gene ID: | 65972 |
Gene Name: | interferon gamma inducible protein 30 |
Gene Aliases: |
GILT, IP30 |
Location: |
Chr.8:70762773-70766663 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifi30 | NM_023065.3 | NP_075552.2 | ||
| AF290973.1 | AAG00598.1 | |||
| AF309649.1 | AAG34681.1 | |||
| AK077725.1 | ||||
| AK156914.1 | BAE33895.1 | |||
| BC054852.1 | AAH54852.1 | |||
| BE951500.1 |
More Information
Set Membership: |
|

Genomic Map