Assay Details
Target Gene Details
Entrez Gene ID: | 66916 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7 |
Gene Aliases: |
1110002H15Rik |
Location: |
Chr.8:83566758-83571623 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufb7 | NM_025843.3 | 1 | 121 | NP_080119.1 |
| AK002678.1 | 1 | 119 | BAB22280.1 | |
| AK003300.1 | 1 | 122 | BAB22701.1 | |
| BC034787.1 | 1 | 100 | AAH34787.1 |
More Information
Set Membership: |
|

Genomic Map