Assay Details
Target Gene Details
Entrez Gene ID: | 66959 |
Gene Name: | dual specificity phosphatase 26 (putative) |
Gene Aliases: |
2310043K02Rik, Skrp3 |
Location: |
Chr.8:31089662-31097047 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp26 | NM_025869.3 | 4 | 782 | NP_080145.1 |
| AB104416.1 | 2 | 572 | BAD91016.1 | |
| AK007513.1 | 2 | 152 | ||
| AK009781.1 | 4 | 783 | BAB26501.2 | |
| BC018204.1 | 2 | 572 | AAH18204.1 |

Genomic Map