Assay Details
Target Gene Details
Entrez Gene ID: | 52815 |
Gene Name: | lactate dehydrogenase D |
Gene Aliases: |
4733401P21Rik, D8Bwg1320e |
Location: |
Chr.8:111626271-111630322 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ldhd | NM_027570.3 | NP_081846.3 | ||
| AK014649.1 | BAB29488.1 | |||
| AY092768.1 | AAM50323.1 | |||
| BC039155.1 | AAH39155.1 | |||
| BC055443.1 | AAH55443.1 |
More Information
Set Membership: |
|

Genomic Map