Assay Details
Target Gene Details
Entrez Gene ID: | 13215 |
Gene Name: | defensin beta 2 |
Gene Aliases: |
BD-2 |
Location: |
Chr.8:21839926-21843482 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Defb2 | NM_010030.1 | 2 | 204 | NP_034160.1 |
| AJ011800.1 | 2 | 204 | CAB42815.1 | |
| BC109129.1 | 2 | 192 | AAI09130.1 | |
| BC109130.1 | 2 | 192 | AAI09131.1 |
More Information
Set Membership: |
|

Genomic Map