Assay Details
Target Gene Details
Entrez Gene ID: | 104394 |
Gene Name: | E2F transcription factor 4 |
Gene Aliases: |
2010111M04Rik, AI427446 |
Location: |
Chr.8:105297663-105305370 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2f4 | NM_148952.1 | NP_683754.1 | ||
| AK145950.1 | BAE26778.1 | |||
| AK157028.1 | BAE33937.1 | |||
| BC023859.1 | AAH23859.1 | |||
| BC026649.1 | AAH26649.1 | |||
| BC027048.1 | AAH27048.1 |
More Information
Set Membership: |
|

Genomic Map