Assay Details
Target Gene Details
Entrez Gene ID: | 66590 |
Gene Name: | phenylalanyl-tRNA synthetase, alpha subunit |
Gene Aliases: |
0610012A19Rik, Farsla |
Location: |
Chr.8:84856972-84869257 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Farsa | NM_025648.3 | NP_079924.2 | ||
| AK002577.1 | ||||
| AK016506.1 | ||||
| AK031697.1 | BAC27519.1 | |||
| AK044637.1 | BAC32014.1 | |||
| AK084031.1 | BAC39103.1 | |||
| AK154629.1 | BAE32726.1 | |||
| AK171872.1 | ||||
| BB853717.1 | ||||
| BC006862.1 | AAH06862.1 | |||
| BC013533.1 | AAH13533.1 | |||
| CT010350.1 | CAJ18558.1 |
More Information
Set Membership: |
|

Genomic Map