Assay Details
Target Gene Details
Entrez Gene ID: | 81007 |
Gene Name: | defensin beta 5 |
Gene Aliases: |
BD-5 |
Location: |
Chr.8:19247592-19250828 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Defb5 | NM_030734.2 | NP_109659.2 |
More Information
Set Membership: |
|

Genomic Map