Assay Details
Target Gene Details
Entrez Gene ID: | 68533 |
Gene Name: | M phase phosphoprotein 6 |
Gene Aliases: |
1110001M01Rik, AA536809, C86426 |
Location: |
Chr.8:117791642-117801933 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mphosph6 | NM_026758.3 | NP_081034.1 | ||
| AK003245.1 | BAB22664.1 | |||
| BC008161.1 | AAH08161.1 |

Genomic Map