Assay Details
Target Gene Details
Entrez Gene ID: | 14894 |
Gene Name: | cilia and flagella associated protein 20 |
Gene Aliases: |
2600014O15Rik, AL024127, Gtl3, T10-2A2 |
Location: |
Chr.8:95420249-95434869 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfap20 | NM_008187.2 | 4 | 710 | NP_032213.2 |
| AK011217.1 | 4 | 654 | ||
| AK088713.1 | 4 | 614 | BAC40521.1 | |
| AK172603.1 | 4 | 686 | BAE43087.1 | |
| BC119023.1 | 4 | 606 | ||
| BC119025.1 | 4 | 606 | ||
| CB195345.1 | 1 | 132 | ||
| Z54179.1 | 4 | 633 | CAA90889.1 |

Genomic Map