Assay Details
Target Gene Details
Entrez Gene ID: | 83436 |
Gene Name: | pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2 |
Gene Aliases: |
6430512N22Rik, AI851530, AU041791, TAPP2, W91773 |
Location: |
Chr.8:25039144-25104917 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekha2 |

Genomic Map