Assay Details
Target Gene Details
Entrez Gene ID: | 70359 |
Gene Name: | GTP binding protein 3 |
Gene Aliases: |
2410009F13Rik, AI607903 |
Location: |
Chr.8:71487870-71499623 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtpbp3 | NM_032544.3 | NP_115933.2 | ||
| AF361482.2 | AAK35216.2 | |||
| AK046330.1 | ||||
| BC092042.1 | AAH92042.1 | |||
| BC138560.1 |
More Information
Set Membership: |
|

Genomic Map