Assay Details
Target Gene Details
Entrez Gene ID: | 14934 |
Gene Name: | glycophorin A |
Gene Aliases: |
AI853584, CD235a, GPA |
Location: |
Chr.8:80493981-80510785 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gypa | NM_010369.3 | NP_034499.3 | ||
| AK145998.1 | BAE26819.1 | |||
| AK157851.1 | BAE34230.1 | |||
| AK162614.1 | BAE36990.1 | |||
| BC148192.1 |

Genomic Map