Assay Details
Target Gene Details
Entrez Gene ID: | 57437 |
Gene Name: | golgi autoantigen, golgin subfamily a, 7 |
Gene Aliases: |
AB041568, C130038N16Rik, GCP16, GOLGA3AP1, HSPC041, R75586 |
Location: |
Chr.8:23241326-23257183 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golga7 | NM_001042484.1 | NP_001035949.1 | ||
| NM_020585.2 | NP_065610.2 | |||
| AB041568.1 | BAA95052.1 | |||
| AK048175.1 | BAC33266.1 | |||
| AK077582.1 | BAC36875.1 | |||
| AK089856.1 | BAC40977.1 | |||
| AK150462.1 | BAE29580.1 | |||
| AK150696.1 | BAE29775.1 | |||
| AK151353.1 | BAE30329.1 | |||
| AK152520.1 | BAE31280.1 | |||
| AK152606.1 | BAE31353.1 | |||
| AK153213.1 | BAE31811.1 | |||
| AK170129.1 | BAE41583.1 | |||
| BC016894.1 | AAH16894.1 | |||
| BC093518.1 | AAH93518.1 | |||
| BC103796.1 | AAI03797.1 | |||
| BY330336.1 |

Genomic Map