Assay Details
Target Gene Details
Entrez Gene ID: | 78849 |
Gene Name: | RIKEN cDNA B430010I23 gene |
Gene Aliases: |
- |
Location: |
Chr.8:41017468-41024216 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B430010I23Rik | NR_015457.1 | 3 | 570 | |
| AK033372.1 | 3 | 570 | ||
| AK046663.1 | 1 | 974 | ||
| AK140364.1 | 2 | 154 |
Target Gene Details
Entrez Gene ID: | 102103 |
Gene Name: | mitochondrial tumor suppressor 1 |
Gene Aliases: |
AI481402, ATBP135, Atip1, B430010I23Rik, B430305I03Rik, C85752, Cctsg1-440, MD44, MTSG1, mKIAA1288 |
Location: |
Chr.8:40990912-41133983 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtus1 | NM_001005863.2 | 6 | 2998 | NP_001005863.1 |
| NM_001005864.3 | 4 | 604 | NP_001005864.1 | |
| NM_001005865.3 | 2 | 617 | NP_001005865.2 | |
| NM_001286413.1 | 6 | 2880 | NP_001273342.1 | |
| AF173380.1 | 2 | 456 | AAD49746.1 | |
| AF493235.1 | 2 | 521 | AAQ06609.1 | |
| AK030510.1 | 2 | 581 | BAC26996.1 | |
| AK031693.1 | 2 | 551 | BAC27517.1 | |
| AK043214.1 | 4 | 591 | BAC31495.1 | |
| AK129324.1 | 2 | 603 | BAC98134.1 | |
| AY246699.1 | 2 | 279 | AAO88908.1 | |
| AY626781.1 | 2 | 614 | AAT45892.1 | |
| AY626782.1 | 4 | 602 | AAT45893.1 | |
| AY626783.1 | 5 | 2820 | AAT45894.1 | |
| BB647317.1 | ||||
| BC041777.1 | 5 | 1663 | AAH41777.1 | |
| BC042206.1 | 5 | 1663 | AAH42206.1 | |
| BC043321.1 | 4 | 568 | AAH43321.1 | |
| BC089009.1 | 6 | 2880 | AAH89009.1 | |
| BE653450.1 | ||||
| BY726879.1 | 2 | 597 |

Genomic Map