Assay Details
Target Gene Details
Entrez Gene ID: | 70984 |
Gene Name: | RIKEN cDNA 4931406C07 gene |
Gene Aliases: |
- |
Location: |
Chr.9:15283337-15306448 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4931406C07Rik | NM_001199484.1 | 2 | 329 | NP_001186413.1 |
NM_001199485.1 | 2 | 225 | NP_001186414.1 | |
NM_133732.3 | 1 | 188 | NP_598493.1 | |
AK016432.1 | 2 | 188 | ||
AK088123.1 | 2 | 189 | BAC40160.1 | |
AK133192.1 | 2 | 330 | ||
AK133768.1 | 2 | 198 | ||
AK135434.1 | 2 | 197 | ||
AK147073.1 | 2 | 199 | BAE27654.1 | |
BC011540.1 | 1 | 91 | AAH11540.1 | |
BC016078.1 | 1 | 137 | AAH16078.1 | |
BY118027.1 | 2 | 226 | ||
CT010296.1 | CAJ18504.1 |
