Assay Details
Target Gene Details
Entrez Gene ID: | 76900 |
Gene Name: | single stranded DNA binding protein 4 |
Gene Aliases: |
1210002E11Rik, AW743380, Sspb4 |
Location: |
Chr.8:70597485-70608380 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ssbp4 | NM_133772.2 | NP_598533.2 | ||
| AK004835.1 | ||||
| AK154336.1 | BAE32522.1 | |||
| AK157387.1 | 10 | 834 | ||
| AK198135.1 | ||||
| AK212417.1 | ||||
| BC002150.1 | AAH02150.1 | |||
| CX237617.1 |
More Information
Set Membership: |
|

Genomic Map