Assay Details
Target Gene Details
Entrez Gene ID: | 71910 |
Gene Name: | phosphatidic acid phosphatase type 2 domain containing 1B |
Gene Aliases: |
1810019D05Rik, 2310022A04Rik, AW045471, Ppapdc1 |
Location: |
Chr.8:25720043-25724887 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppapdc1b | NM_001293703.1 | NP_001280632.1 | ||
| NM_028000.1 | NP_082276.1 | |||
| NR_121642.1 | ||||
| NR_121643.1 | ||||
| AA184457.1 | ||||
| AK007542.1 | 3 | 438 | ||
| AK009455.1 | 5 | 444 | ||
| AK144593.1 | BAE25955.1 | |||
| BB857717.1 | ||||
| BC023111.1 | ||||
| BC062173.1 | AAH62173.1 | |||
| BC089556.1 | AAH89556.1 | |||
| BC099489.1 | AAH99489.1 | |||
| BE646843.1 | ||||
| DV661598.1 |
More Information
Set Membership: |
|

Genomic Map