Assay Details
Target Gene Details
Entrez Gene ID: | 66952 |
Gene Name: | RIKEN cDNA 2310030G06 gene |
Gene Aliases: |
2610024D04Rik, AV017521 |
Location: |
Chr.9:50739691-50746521 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2310030G06Rik | NM_025865.2 | NP_080141.1 | ||
| AK007611.1 | BAB25134.1 | |||
| AK007929.1 | BAB25356.1 | |||
| AK009541.1 | BAB26348.1 | |||
| BC027409.1 | AAH27409.1 |

Genomic Map