Assay Details
Target Gene Details
Entrez Gene ID: | 622675 |
Gene Name: | zinc finger protein 827 |
Gene Aliases: |
2810449M09Rik, D630040G17Rik, Znf827 |
Location: |
Chr.8:79027983-79193768 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp827 | NM_001294279.1 | 6 | 2297 | NP_001281208.1 |
| NM_178267.3 | 6 | 2297 | NP_839998.2 | |
| AK154905.1 | 6 | 2150 | ||
| BC151183.1 | 6 | 2182 | ||
| BU055006.1 | 2 | 307 | ||
| CN461771.1 | 3 | 751 |
More Information
Set Membership: |
|

Genomic Map