Assay Details
Target Gene Details
Entrez Gene ID: | 622675 |
Gene Name: | zinc finger protein 827 |
Gene Aliases: |
2810449M09Rik, D630040G17Rik, Znf827 |
Location: |
Chr.8:79027983-79193768 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp827 | NM_001294279.1 | 9 | 2691 | NP_001281208.1 |
| NM_178267.3 | 9 | 2691 | NP_839998.2 | |
| AK052736.1 | 4 | 255 | BAC35124.1 | |
| AK086577.1 | 1 | 90 | ||
| AK154905.1 | 9 | 2544 | ||
| BC088739.1 | 4 | 327 | AAH88739.1 | |
| BC151183.1 | 9 | 2576 | ||
| BU055006.1 |
More Information
Set Membership: |
|

Genomic Map