Assay Details
Target Gene Details
Entrez Gene ID: | 320158 |
Gene Name: | zinc finger, matrin type 4 |
Gene Aliases: |
9630048M01Rik |
Location: |
Chr.8:23669547-24063117 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zmat4 | NM_001277239.1 | 5 | 455 | NP_001264168.1 |
| NM_177086.4 | 4 | 414 | NP_796060.1 | |
| AK036254.1 | 4 | 413 | BAC29363.1 | |
| AK134793.1 | 4 | 413 | BAE22287.1 | |
| AK140413.1 | 4 | 415 | BAE24377.1 | |
| BC065110.1 | 4 | 402 | AAH65110.1 | |
| BC079657.1 | 5 | 442 | AAH79657.1 |

Genomic Map