Assay Details
Target Gene Details
Entrez Gene ID: | 234695 |
Gene Name: | RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing |
Gene Aliases: |
CG1399-PB, D130029J02Rik, Gm585 |
Location: |
Chr.8:105686010-105698195 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rltpr | NM_001033320.2 | NP_001028492.1 | ||
| AK031012.1 | BAE20471.1 | |||
| BC115830.1 | ||||
| HF678090.1 |

Genomic Map