Assay Details
Target Gene Details
Entrez Gene ID: | 320165 |
Gene Name: | transforming, acidic coiled-coil containing protein 1 |
Gene Aliases: |
4833447E04Rik, AA960202, B230378H13Rik |
Location: |
Chr.8:25154552-25241585 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tacc1 | NM_177089.5 | NP_796063.3 | ||
| NM_199323.3 | NP_955355.1 | |||
| AK158441.1 | ||||
| AY177412.1 | AAO53447.1 | |||
| AY177413.1 | AAO53448.1 | |||
| BC125389.1 | ||||
| BC145709.1 |

Genomic Map