Assay Details
Target Gene Details
Entrez Gene ID: | 21341 |
Gene Name: | TATA box binding protein (Tbp)-associated factor, RNA polymerase I, C |
Gene Aliases: |
Tafi95, mTAFI95 |
Location: |
Chr.8:119597974-119605240 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Taf1c | NM_021441.2 | NP_067416.2 | ||
| AK030522.1 | ||||
| AK052250.1 | ||||
| AK087534.1 | BAC39916.1 | |||
| AK154846.1 | BAE32872.1 | |||
| AK156718.1 | BAE33818.1 | |||
| BC058383.1 | AAH58383.1 | |||
| Y09974.1 | CAA71093.1 |
More Information
Set Membership: |
|

Genomic Map