Assay Details
Target Gene Details
Entrez Gene ID: | 69742 |
Gene Name: | TM2 domain containing 2 |
Gene Aliases: |
2410018G23Rik, Blp1 |
Location: |
Chr.8:25017211-25023260 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tm2d2 | NM_027194.3 | NP_081470.2 | ||
| AK010545.1 | BAB27019.1 | |||
| AK045768.1 | BAC32484.1 | |||
| AK080854.1 | ||||
| AK083654.1 | BAC38985.1 | |||
| AK085975.1 | BAC39582.1 | |||
| AK086015.1 | ||||
| AK086059.1 | ||||
| AK088246.1 | ||||
| AK149407.1 | BAE28855.1 | |||
| AK160039.1 | BAE35581.1 | |||
| AK166012.1 | BAE38519.1 | |||
| AK206565.1 | ||||
| BC026789.1 | AAH26789.1 |

Genomic Map