Assay Details
Target Gene Details
Entrez Gene ID: | 234129 |
Gene Name: | transmembrane phosphatase with tensin homology |
Gene Aliases: |
Pten2 |
Location: |
Chr.8:22283441-22371418 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpte | NM_199257.2 | NP_954866.2 | ||
| AJ311311.1 | CAC44243.1 | |||
| AJ311312.1 | CAC44244.1 | |||
| AJ311313.1 | CAC44245.1 | |||
| BC107329.2 | AAI07330.1 |
More Information
Set Membership: |
|

Genomic Map