Assay Details
Target Gene Details
Entrez Gene ID: | 83436 |
Gene Name: | pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2 |
Gene Aliases: |
6430512N22Rik, AI851530, AU041791, TAPP2, W91773 |
Location: |
Chr.8:25039144-25104917 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekha2 | NM_031257.3 | NP_112547.1 | ||
| AF286161.1 | AAG15198.1 | |||
| AF418551.1 | AAL57436.1 | |||
| AK032325.1 | ||||
| AK042324.1 | BAC31224.2 | |||
| AK078212.1 | BAC37176.1 | |||
| AK088395.1 | BAC40325.1 | |||
| AK153117.1 | ||||
| BC010215.1 | AAH10215.1 | |||
| BI730323.1 |

Genomic Map