Assay Details
Target Gene Details
Entrez Gene ID: | 77733 |
Gene Name: | ring finger protein 170 |
Gene Aliases: |
6720407G21Rik, AI481227 |
Location: |
Chr.8:26119368-26143874 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf170 | NM_029965.2 | NP_084241.1 | ||
| AK020104.1 | ||||
| AK036051.1 | BAC29287.1 | |||
| AK042063.1 | BAC31147.1 | |||
| AK080591.1 | BAC37951.1 | |||
| AK152760.1 | BAE31474.1 | |||
| AK153658.1 | ||||
| AK162554.1 | ||||
| BC031383.1 | AAH31383.1 | |||
| BC114210.1 | AAI14211.1 | |||
| BC138930.1 | ||||
| BC145718.1 |

Genomic Map