Assay Details
Target Gene Details
Entrez Gene ID: | 75767 |
Gene Name: | RAB11 family interacting protein 1 (class I) |
Gene Aliases: |
2010200K21Rik, 4833414G05Rik, Rcp |
Location: |
Chr.8:27138773-27174646 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab11fip1 | NM_001080813.2 | 5 | 3297 | NP_001074282.1 |
| NM_029423.2 | 4 | 1734 | NP_083699.2 | |
| AK014696.1 | 4 | 1716 | BAB29507.1 | |
| AK030769.1 | 4 | 1737 | ||
| AK086797.1 | 4 | 1732 | ||
| AK131172.1 | 2 | 228 | BAD21422.1 | |
| AK151024.1 | 4 | 1735 | BAE30042.1 | |
| BC125400.1 | 4 | 1719 | ||
| BC132094.1 | 4 | 1719 |

Genomic Map