Assay Details
Target Gene Details
Entrez Gene ID: | 30927 |
Gene Name: | snail family zinc finger 3 |
Gene Aliases: |
AI643946, Smuc, Zfp293 |
Location: |
Chr.8:122454204-122460927 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snai3 | NM_013914.2 | NP_038942.1 | ||
| AF133714.1 | AAF22956.1 | |||
| AF195655.1 | AAQ13827.1 | |||
| AK042234.1 | BAC31201.1 | |||
| AK089285.1 | BAC40828.1 | |||
| AK153633.1 | BAE32128.1 | |||
| BC100724.1 | AAI00725.1 | |||
| BC100726.1 | AAI00727.1 | |||
| BC100727.1 | AAI00728.1 |
More Information
Set Membership: |
|

Genomic Map