Assay Details
Target Gene Details
Entrez Gene ID: | 234371 |
Gene Name: | transmembrane protein 161A |
Gene Aliases: |
AI428876, BB161850, BC021367 |
Location: |
Chr.8:70172329-70183681 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem161a |

Genomic Map