Assay Details
Target Gene Details
Entrez Gene ID: | 234371 |
Gene Name: | transmembrane protein 161A |
Gene Aliases: |
AI428876, BB161850, BC021367 |
Location: |
Chr.8:70172329-70183681 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem161a | NM_001293797.1 | NP_001280726.1 | ||
| NM_145597.4 | NP_663572.1 | |||
| NR_122028.1 | ||||
| AK032711.1 | BAC27995.1 | |||
| AK049979.1 | BAC34016.1 | |||
| AK082758.1 | ||||
| AK082916.1 | BAC38686.1 | |||
| AK088569.1 | BAC40428.1 | |||
| AK147962.1 | BAE28253.1 | |||
| AK167517.1 | ||||
| AK180152.1 | ||||
| AK198557.1 | ||||
| AK215127.1 | ||||
| BC021367.1 | AAH21367.1 |

Genomic Map