Assay Details
Target Gene Details
Entrez Gene ID: | 76808 |
Gene Name: | ribosomal protein L18A |
Gene Aliases: |
2510019J09Rik |
Location: |
Chr.8:70894722-70897443 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl18a | NM_029751.4 | NP_084027.1 | ||
| AK010983.1 | BAB27304.1 | |||
| AK146543.1 | BAE27248.1 | |||
| AK150820.1 | ||||
| AK152611.1 | ||||
| AK161942.1 | BAE36645.1 | |||
| AK162047.1 | BAE36696.1 | |||
| AK168258.1 | BAE40206.1 | |||
| BC024609.1 | ||||
| BC037146.1 | AAH37146.1 | |||
| CJ042325.1 |
More Information
Set Membership: |
|

Genomic Map