Assay Details
Target Gene Details
Entrez Gene ID: | 66817 |
Gene Name: | transmembrane protein 170 |
Gene Aliases: |
9030409E16Rik, AA403674, D8Bwg1414e, Tmem170a |
Location: |
Chr.8:111863038-111876863 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem170 | NM_025781.2 | 1 | 45 | NP_080057.1 |
| AK007926.1 | 1 | 90 | ||
| AK018496.1 | 1 | 46 | BAB31240.1 | |
| BC139377.1 | 1 | 120 | ||
| BC145964.1 | 1 | 120 |

Genomic Map