Assay Details
Target Gene Details
Entrez Gene ID: | 56513 |
Gene Name: | par-6 family cell polarity regulator alpha |
Gene Aliases: |
0710008C04Rik, 2610010A15Rik, PAR-6A, PAR6alpha, Par-6, Par6, Par6c, TAX40, Tip-40 |
Location: |
Chr.8:105701148-105703494 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pard6a | NM_001047436.2 | 1 | 37 | NP_001040901.1 |
| NM_001286344.1 | 1 | 37 | NP_001273273.1 | |
| AK019153.1 | 1 | 35 | ||
| BY099545.1 | 1 | 37 | ||
| CK618363.1 | ||||
| CX206739.1 | ||||
| CX220562.1 | 1 | 31 |
More Information
Set Membership: |
|

Genomic Map