Assay Details
Target Gene Details
Entrez Gene ID: | 56437 |
Gene Name: | Ras-related associated with diabetes |
Gene Aliases: |
REM3, Rad |
Location: |
Chr.8:104628066-104631367 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrad | NM_019662.2 | NP_062636.2 | ||
| AF084466.1 | AAC33133.1 | |||
| AK077271.1 | ||||
| AK087473.1 | ||||
| AK159550.1 | BAE35175.1 | |||
| BC057138.1 | AAH57138.1 |

Genomic Map