Assay Details
Target Gene Details
Entrez Gene ID: | 56437 |
Gene Name: | Ras-related associated with diabetes |
Gene Aliases: |
REM3, Rad |
Location: |
Chr.8:104628066-104631367 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrad | NM_019662.2 | 4 | 815 | NP_062636.2 |
| AF084466.1 | 3 | 558 | AAC33133.1 | |
| AK077271.1 | 4 | 813 | ||
| AK087473.1 | 4 | 788 | ||
| AK159550.1 | 3 | 598 | BAE35175.1 | |
| BC057138.1 | 4 | 794 | AAH57138.1 |

Genomic Map