Assay Details
Target Gene Details
Entrez Gene ID: | 74166 |
Gene Name: | transmembrane protein 38A |
Gene Aliases: |
1110001E17Rik, AI413399, TRIC-A, mg33a |
Location: |
Chr.8:72571998-72587284 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem38a | NM_144534.1 | NP_653117.1 | ||
| AB261158.1 | ||||
| AK157969.1 | BAE34290.1 | |||
| AK165818.1 | BAE38393.1 | |||
| BC014728.1 | AAH14728.1 | |||
| BC024788.1 | AAH24788.1 | |||
| EF988666.1 |
More Information
Set Membership: |
|

Genomic Map