Assay Details
Target Gene Details
Entrez Gene ID: | 56513 |
Gene Name: | par-6 family cell polarity regulator alpha |
Gene Aliases: |
0710008C04Rik, 2610010A15Rik, PAR-6A, PAR6alpha, Par-6, Par6, Par6c, TAX40, Tip-40 |
Location: |
Chr.8:105701148-105703494 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pard6a | NM_001047435.2 | 2 | 268 | NP_001040900.1 |
| NM_001047436.2 | 2 | 281 | NP_001040901.1 | |
| NM_001286344.1 | 2 | 250 | NP_001273273.1 | |
| NM_001286345.1 | NP_001273274.1 | |||
| NM_019695.3 | 2 | 268 | NP_062669.2 | |
| AF070970.1 | 2 | 264 | AAD15928.1 | |
| AK003030.1 | 2 | 238 | ||
| AK158848.1 | 2 | 266 | BAE34693.1 | |
| AK193466.1 | ||||
| BC049593.1 | 2 | 245 | AAH49593.1 | |
| BC061114.1 | AAH61114.1 | |||
| BG806503.1 | 2 | 193 | ||
| BY099545.1 | 2 | 250 | ||
| BY280124.1 | 2 | 266 | ||
| CK618363.1 | 2 | 224 | ||
| CX206739.1 | 2 | 231 | ||
| CX220562.1 | 2 | 275 |
More Information
Set Membership: |
|

Genomic Map