Assay Details
Target Gene Details
Entrez Gene ID: | 234214 |
Gene Name: | sorbin and SH3 domain containing 2 |
Gene Aliases: |
2010203O03Rik, 9430041O17Rik, A530071H08, Argbp2, mKIAA0777 |
Location: |
Chr.8:45507629-45827906 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 20 - Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sorbs2 | NM_001205219.1 | NP_001192148.1 | ||
| NM_172752.4 | NP_766340.4 | |||
| AK050951.1 | ||||
| AK083429.1 | BAC38913.1 | |||
| AK122369.1 | BAC65651.1 | |||
| AK140498.1 | BAE24407.1 | |||
| BC039163.1 | AAH39163.1 | |||
| BC151038.1 | ||||
| BC157944.1 | ||||
| BC172140.1 |
Target Gene Details
Entrez Gene ID: | 319940 |
Gene Name: | sorbin and SH3 domain containing 2, opposite strand |
Gene Aliases: |
D330022K07Rik |
Location: |
Chr.8:45723325-45819297 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sorbs2os | NR_045739.1 | |||
| AK084672.1 |
More Information
Set Membership: |
|

Genomic Map