Assay Details
Target Gene Details
Entrez Gene ID: | 234214 |
Gene Name: | sorbin and SH3 domain containing 2 |
Gene Aliases: |
2010203O03Rik, 9430041O17Rik, A530071H08, Argbp2, mKIAA0777 |
Location: |
Chr.8:45507629-45827906 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sorbs2 | NM_001205219.1 | NP_001192148.1 | ||
NM_172752.4 | NP_766340.4 | |||
AK050951.1 | ||||
AK079130.1 | BAC37554.1 | |||
AK083429.1 | BAC38913.1 | |||
AK139388.1 | BAE23988.1 | |||
AK140498.1 | BAE24407.1 | |||
AK171632.1 | ||||
BC039163.1 | AAH39163.1 | |||
BC151038.1 | ||||
BC157944.1 | ||||
BC172140.1 | ||||
CV883960.1 |
Target Gene Details
Entrez Gene ID: | 319940 |
Gene Name: | sorbin and SH3 domain containing 2, opposite strand |
Gene Aliases: |
D330022K07Rik |
Location: |
Chr.8:45723325-45819297 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sorbs2os | NR_045739.1 | |||
AK084672.1 |
