Assay Details
Target Gene Details
Entrez Gene ID: | 18408 |
Gene Name: | solute carrier family 25 (mitochondrial carrier ornithine transporter), member 15 |
Gene Aliases: |
D630044L02Rik, Ornt1 |
Location: |
Chr.8:22375550-22398621 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a15 | NM_181325.4 | NP_851842.1 | ||
| AF133914.1 | AAD45252.1 | |||
| AK052758.1 | BAC35136.1 | |||
| AK081243.1 | 4 | 598 | ||
| AK142223.1 | BAE24984.1 | |||
| AK145966.1 | BAE26791.1 | |||
| AK154988.1 | ||||
| BC061143.1 | AAH61143.1 |
More Information
Set Membership: |
|

Genomic Map