Assay Details
Target Gene Details
Entrez Gene ID: | 18408 |
Gene Name: | solute carrier family 25 (mitochondrial carrier ornithine transporter), member 15 |
Gene Aliases: |
D630044L02Rik, Ornt1 |
Location: |
Chr.8:22375550-22398621 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a15 | NM_181325.4 | 5 | 740 | NP_851842.1 |
| AF133914.1 | 5 | 588 | AAD45252.1 | |
| AK052758.1 | 5 | 672 | BAC35136.1 | |
| AK081243.1 | 4 | 2863 | ||
| AK142223.1 | 5 | 610 | BAE24984.1 | |
| AK145966.1 | 5 | 610 | BAE26791.1 | |
| AK154988.1 | 5 | 716 | ||
| BC061143.1 | 5 | 381 | AAH61143.1 |
More Information
Set Membership: |
|

Genomic Map