Assay Details
Target Gene Details
Entrez Gene ID: | 76687 |
Gene Name: | signal peptidase complex subunit 3 homolog (S. cerevisiae) |
Gene Aliases: |
1810011E08Rik |
Location: |
Chr.8:54520433-54530739 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spcs3 | NM_029701.1 | NP_083977.1 | ||
| AK007432.1 | BAB25035.1 | |||
| AK028314.1 | BAC25873.1 | |||
| AK048051.1 | BAC33224.1 | |||
| BC054817.1 | AAH54817.1 | |||
| BY012914.1 |
More Information
Set Membership: |
|

Genomic Map