Assay Details
Target Gene Details
Entrez Gene ID: | 67023 |
Gene Name: | unconventional SNARE in the ER 1 homolog (S. cerevisiae) |
Gene Aliases: |
2010315L10Rik, 5730403H22Rik, AV002165, D12, Ed2, Q-snare, p31 |
Location: |
Chr.8:71366848-71369732 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Use1 | NM_001145780.1 | NP_001139252.1 | ||
| NM_025917.4 | NP_080193.1 | |||
| AF353245.1 | AAK26661.1 | |||
| AK003257.1 | ||||
| AK003385.1 | ||||
| AK008573.1 | BAB25752.1 | |||
| AK008661.1 | ||||
| AK012490.1 | BAB28275.1 | |||
| AK017484.1 | BAB30767.1 | |||
| AK160726.1 | BAE35971.1 | |||
| AK165639.1 | ||||
| AY842453.1 | AAW32097.1 | |||
| BC025086.1 | ||||
| BC038519.1 | AAH38519.1 | |||
| BC075695.1 | AAH75695.1 |

Genomic Map