Assay Details
Target Gene Details
Entrez Gene ID: | 22152 |
Gene Name: | tubulin, beta 3 class III |
Gene Aliases: |
3200002H15Rik, M(beta)3, M(beta)6 |
Location: |
Chr.8:123411564-123422010 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tubb3 | NM_023279.2 | NP_075768.1 | ||
| AF312873.1 | AAG26010.1 | |||
| AK012528.1 | BAB28299.1 | |||
| AK051298.1 | BAC34596.1 | |||
| AK082942.1 | ||||
| AK149014.1 | BAE28719.1 | |||
| BC031357.1 | AAH31357.1 | |||
| BC088749.1 | AAH88749.1 |
More Information
Set Membership: |
|

Genomic Map